b'SeparateTransferDetectCasting multiple gels because of leaks? Pour-your-own gels with a 100% leak-free system* Features of the SureCast Gel Handcast System include:Pour your own polyacrylamide mini gels easily and Leak-free designgels that are more usable, for less confi dently with the Invitrogen SureCast Gel Handcastwasted timeSystem. The SureCast system is fully compatible with our Mini Gel Tank. Use Invitrogen SureCast handcasting Superior glass plate durabilityup to 20 times more reagents as well as other popular polyacrylamide geldurable compared to other suppliers platescasting reagents. Unique tilt featurehelps minimize spillage when pouring acrylamide solutions Simple assembly of casting componentsuses a * Terms and conditions apply. For complete details, go to thermofi sher.com/surecastterms. single-motion, load-and-lock mechanismWelcome packsSureCast Gel Handcast Bundle A SureCast Gel Handcast Bundle BThe Invitrogen SureCast Gel Handcast Bundle A includes all theThe Invitrogen SureCast Gel Handcast Bundle B includes all the hardware and reagents to get started: hardware to get started:2 SureCast Gel Handcast Stations 2 SureCast Gel Handcast Stations2 SureCast Glass Plate Sets (2 front, 2 back) 2 SureCast Glass Plate Sets (2 front, 2 back)2 SureCast Sealing Pads (attached to handcast stations) 2 SureCast Sealing Pads (attached to handcast stations)10 SureCast Gel Spacers 10 SureCast Gel Spacers1 10-Well Multi-Use Tool 1 10-Well Multi-Use Tool1 12-Well Multi-Use Tool 1 12-Well Multi-Use Tool1 15-Well Multi-Use Tool 1 15-Well Multi-Use Tool6 Gel Combs (2 each: 10-, 12-, 15-well) 6 Gel Combs (2 each: 10-, 12-, 15-well)1 SureCast Stacking Buff er 2-pack1 SureCast Resolving Buff er 2-pack1 SureCast APS (25 g)1 SureCast TEMED (30 mL)1 SureCast Acrylamide Solution, 40% (450 mL) Find out more at thermofi sher.com/surecast36'