b'SeparateTransferDetectRun or transfer up to 2 Invitrogen midi gels in the same tank Electrophoresis chamber systemsThe SureLock Tandem Midi Gel Tank was designed for easy and consistent vertical protein gel electrophoresis of 1 or 2 Invitrogen midi gels. When paired with the Invitrogen SureLock Midi Transfer Module, this tank performs efficient, 30-minute, room-temperature, wet protein transfers. Separate chambers make it easy to run just one gel or transfer, allowing you to save on buff er and limit methanol waste and disposal costs. Save the time, planning, and mess required with other systems to pre-chill buff ers, freeze ice packs, or prepare ice baths. Convenient, precut membranes and fi lter papers are available to speed up your work.The SureLock Tandem Midi Gel Tank is available separately or in a variety of product bundles for built-in savings. Welcome packs are available with the SureLock Tandem Midi Blot Module and PVDF or nitrocellulose membranes, and Invitrogen SureLock Tandem Transfer Tray. Simultaneous electrophoresis of up to four midi gelsXCell4 SureLock Midi-Cell systemThe Invitrogen XCell4SureLock Midi-Cell system allows simultaneous vertical electrophoresis of 14 midi gels, enabling high throughput. It uses unique technology to make electrophoresis easier and more reliable, and is designed to dissipate heat eff ectively and evenly to enable high-resolution results when using Invitrogen midi gels.Learn more at thermofi sher.com/surelocktandem20'