b'SeparateTransferDetectRevolutionary western blot processingno shakers, trays, or timers requiredAutomated iBind Western SystemsInvitrogen iBind Western Systems are automated western blot processing platforms. Simply load primary antibody, secondary antibody, and wash solutions, and then walk away. In less than three hours, the blot is ready for fi naldetection. iBind Western Systems off er: Flexibilitypick the system that matches your throughput; process 1 mini or midi blot, 2 mini blots, or 6 vertically cut strips using the same or diff erent conditionsiBind Western Starter Kit (Cat. No. SLF1000S) Antibody savingsuse up to 80% less Reproducibilityautomated blot processing primary antibody enables improved blot-to-blot consistency Load and gothe system processes solutions using sequential lateral fl ow technology, with no batteries, shakers, trays, or timers requirediBind Western Device iBind Flex Western DeviceMini blot (8 x 8 cm), singleYes YesMini blot (8 x 8 cm), dualNo YesMidi blot (13 x 8 cm) No YesVertically cut strips (up to 6) No YesWatch a video demonstration at thermofi sher.com/ibind12'