b'SeparateTransferDetectExcellent western performance compared to manual blottingStrip 1 Strip 2 Strip 3 Strip 4 Strip 5 Strip 6iBind Flex Western Device Manual (tray) processingFluorescence detection with the iBind Flex Western Device. Comparison of strip blots processed manually (probing and washing steps performed in a tray) vs. with the iBind Flex Western Device. Blots were produced by separating samples on Bolt 412%, 10-well gels with MES SDS running buffer, rapid-dry transfer to nitrocellulose membrane using the iBlot 2 system and then cutting each into 3-lane strips. Get more information about the target proteins and antibodies used at thermofisher.com/ibind. Watch a video demonstration at thermofisher.com/ibind13'