b'SeparateTransferDetectConvenient, reliable western wet transfer Two blot modules fi t into the Mini Gel Tank and theSureLock Tandem Midi Gel Tank. These tanks are designed to make your western transfers simple and easy to perform.Universal module designallows modules to fi t in either chamber of the tank, simplifying the transfer setup Unique gasket sealhelps prevent buff er leakage so there is no mess during setup of your western transfer Half-inch buff er chamberrequires half the typical volume of methanol-based transfer buff ers,reducing hazardous waste and disposal cost (220 mL per mini blot; 300 mL per midi blot) Standard 3060 min transfer protocolaccelerates your western workfl ow so you can get results faster Room-temperature transfereliminate the need to prechill buff ers and the hassle and messiness of ice bathsFind out more at thermofi sher.com/wettransfer40'