b'SeparateTransferDetectUnique chemistries for your unique proteinPrecast gelsPrecast gels offer convenience, speed, andAvailable gel chemistries: consistency. Invitrogen precast gels are Tris-glycine chemistry for broad-range, available in four different chemistries and a widehigh-abundance protein separationvariety of percentages, gradients, sample-well configurations, and formats. Gel formats include Bis-Tris chemistry for broad-range, mini gels or the wider midi gel. The choicelow-abundance protein separation or for of which chemistry to use depends on theapplications requiring high protein integrity, abundance and size of the protein, and yoursuch as posttranslational modification analysis, downstream application. mass spectrometry, or sequencingTris-acetate chemistry for high molecular weight protein separationTricine chemistry for low molecular weight protein separationLoad more with optimal separation Invitrogen Bolt Bis-Tris Plus mini gels are MagicMark standardGST IKK EPHB3 HCK MAPK14 FLT1 DDR2 MagicMark standardGST IKK EPHB3 HCK MAPK14 FLT1 DDR2precast polyacrylamide gels designed for optimal separation of a broad molecular weight range ofproteins under denaturing conditions. Bolt gels are designed to deliver western performance superior to that of Tris-glycinebased gels. Preserved protein integrityneutral-pH formulation minimizes protein modifications,for publication-quality results the first time Bolt Bis-Tris Plus gel Bio-Rad TGX gel High sample-volume capacityWedgeWellBolt Bis-Tris Plus mini gels help provide better western design with up to 60 L sample capacityblotting results. A western blot of a Bolt gel shows clean, sharp protein signals corresponding to only full-length proteins, allows eaiser loading and detection of proteinswhereas a western blot of a Bio-Rad TGX gel shows multiple in very dilute samples or visualization oflow molecular weight degradation products.low-abundance proteins Better band quality and band volume High lot-to-lot consistencycoefficient ofDesigned to deliver sharp, straight bands with variation (CV) of only 2% for R values (migration) higher band volumefExplore protein gel options at thermofisher.com/proteingels8'