b'SeparateTransferDetectRunning or transferring from 1 to 16 gelswe have a power supply that can support your power needsPowerEase power supplies The Invitrogen PowerEase Touch 350W Power Supply brings a high level of convenience to your electrophoresis experiments. With a bright LCD touchscreen interface, you can enter in custom programs, or use the preprogrammed protocols for Invitrogen protein gels and gel transfers. It can run up to 8 midi gels or transfer up to 4 midi gels simultaneously.Each program can include up to20 steps, for precise control over electrophoresis conditions. In addition, the PowerEase Touch 350W Power Supply features: Constant voltage, current, or power settings Built-in timer for walk-away gel electrophoresis Storage for up to100 custom programs with20 steps each Four sets of output jacks that are compatible with most electrophoresis devicesThe Invitrogen PowerEaseTouch 120W Power Supply is designed specifi cally for mini-gel electrophoresis. The straightforward, intuitive interface makes the powering of gel runs a simple and easy process. In addition, the PowerEase 120W Power Supply features: Constant voltage, current, or power settings Built-in timer for walk-away gel electrophoresis Output jacks that are compatible with most electrophoresis devicesPowerEase Touch 120W and 350W Power Supplies are also available in money-saving bundles with a Mini Gel Tank or Invitrogen XCell SureLock Mini-Cell, with or without a blot module, or with an Invitrogen SureLock Tandem Midi Gel Tank, with or without a blot module.Find out more at thermofi sher.com/powerease18'