b'Power Blotter SystemHigh performance semi-dry transfers in less than 10 minutes The Invitrogen Power Blotter and Power Blotter XL Systems are designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 7 to 10 minutes (Figure 6). The Power Blotter features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.Features:Efficienthigh transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wetBoth the Power Blotter System and Power Blotter XL transfer methods (Figure 7) System have an easy-to-use color LCD touchscreen interface with preprogrammed transfer methods forFast7- to 10-minute transfer when used withdifferent numbers and sizes of gels and protein molecular Invitrogen Power Blotter Select Transfer Stacks orweight ranges. The easy-touch programming feature allows Invitrogen Power Blotter 1-Step Transfer Buffer custom transfer settings to be quickly created, saved,Integrated power supplyseamless operationand run. Both systems achieve highly efficient transfer in between control unit and cassette; no external power7 minutes when used with Power Blotter Select Transfer supply necessary Stacks or 10 minutes with Power Blotter 1-Step Transfer Buffer. The instruments are also effective for standardEasy-touch programminguse convenient30- to 60-minute semi-dry transfer protocols based on preprogrammed transfer methods or programtraditional buffers.customized transfer methods Flexibletransfer 12 mini-size or 1 midi-size gel using the Power Blotter Cassette; transfer 14 mini-size or 12 midi-size gels with the Power Blotter Cassette XL Versatilecompatible with Power Blotter Select Transfer Stacks and Invitrogen Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methodsPower Blotter System Power Blotter XL SystemTransfer 12 mini-size gelsTransfer 14 mini-size gelsor 1 midi-size gel or 2 midi-size gelsFigure 6. Power Blotter System formats. Each system includes the Power Blotter Station and either the Power Blotter Cassette or Power Blotter Cassette XL. The station and cassettes are also available separately.11'