b'Nitrocellulose membranes High protein-binding affinity and compatibility with aFeatures:variety of detection methods make nitrocellulose a popular High qualitypure, 100% nitrocellulose membranesmatrix. Protein immobilization on the membrane is thoughtwith high surface area and excellent uniformityto occur by hydrophobic interactions. Use of high saltSelectionavailable in 0.2 m and 0.45 m pore sizes and low methanol concentrations in transfer conditions improves protein immobilization on the membrane,for peptide and protein applications, respectivelyespecially with proteins of higher molecular weights. Convenientavailable in pre-cut sheets, rolls,ready-to-use formats with membrane included, or in We offer a variety of nitrocellulose membranes to supportspecialized device-specific formatsmost of your transfer needs. A comparison of the various offerings can be found in Table 3. High sensitivityprovides high-affinity protein binding, blocks easily, and exhibits very low background in chemiluminescent western blottingTable 3. Thermo Scientific and Invitrogen nitrocellulose membranes.Pore sizeMembrane0.2 m 0.45 msizeMembrane with filter papers Membrane only Membrane with filter papers Membrane onlyMini Nitrocellulose/Filter PaperNitrocellulose Membranes,Nitrocellulose/Filter PaperNitrocellulose Membranes, Sandwiches, 0.2 m, 0.2 m, Sandwiches, 0.45 m, 0.45 m,8.3 x 7.3 cm 8 x 8 cm 8.3 x 7.3 cm 8 x 8 cm(Cat. No. LC2000) (Cat. No. 88024) (Cat. No. LC2001) (Cat. No. 88025)Midi Nitrocellulose/Filter PaperNitrocellulose Membranes,Nitrocellulose/Filter PaperNitrocellulose Membranes, Sandwiches, 0.2 m, 0.2 m, Sandwiches, 0.45 m,0.45 m,8.5 x 13.5 cm 8 x 12 cm 8.5 x 13.5 cm 8 x 12 cm(Cat. No. LC2009) (Cat. No. 77012) (Cat. No. LC2006) (Cat. No. 77010)SureLock Tandem Midi Pre-cutSureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 m,Membranes and Filters, 0.45 m, nitrocellulose, 8.6 x 14 cmnitrocellulose, 8.6 x 14 cm (Cat. No. STM2007) (Cat. No. STM2008)Roll Nitrocellulose Membrane,NANANA 0.45 m,30 cm x 3.5 m(Cat. No. 88018)Learn more atthermofisher.com/membranes17'