b'iBright Imaging Systems Experience an easier time capturing and analyzing data from gels and western blots with Invitrogen iBright Imaging Systems. Designed with a streamlined, intuitive interface and workflows, iBright Imaging Systems are easy to use for researchers of all experience levels. There are three models in the iBright Imaging Systems family: the iBright CL750 Imaging System, the iBright CL1500 Imaging System, and the iBright FL1500 Imaging System. The iBright CL750 Imaging System offers the core essential western blot and gel imaging functions and makes the transition from the darkroom and film easy. The iBright CL1500 Imaging System expands application support and has many of the high-performance specifications of our premier iBright FL1500 model. The iBright FL1500 Imaging System features maximum application support, including fluorescent western blot imaging with up to four fluorescence channels at a time. Features:Push-button, optimized exposureSmartExposure acquisition technology for the rapid determination ofView overview video:optimal exposure times helps minimize the need to repeat exposures to get the desired signal Powerful 9.1 megapixel (MP) cameracapture crystal-clear images with robust imaging potential Advanced automated featuresautomatic sample rotation, automatic zoom, automatic focus, and automatic onboard data analysis provide a smooth imaging experience Five-channel fluorescent blottingmultiplex with the five fluorescence channels of the iBright FL1500 model; capture up to four proteins in a single blot for more meaningful and representative experiments Compliance supportall models offered with 21 CFR Part 11 compliance support software packages to set up and control security, audit, and e-signature settingsCapture your data faster and more easily than ever before.Learn more at thermofisher.com/ibright64'