b'SuperSignal West HisProbe KitThe Thermo Scientific SuperSignal West HisProbe Kit is a chemiluminescent system that uses HisProbe-HRP conjugate to overcome the limitations of anti-histidine antibodies and other detection strategies. HisProbe-HRP conjugate is more specific for polyhistidine tags, reducing background problems. Unlike anti-His antibodies, HisProbe-HRP conjugate can recognize polyhistidine tags independent of adjacent tags.Features: FastHis-tag detection is achieved with a one-step probe incubation that eliminates the need to run a lengthy two-step, primarysecondary antibody reaction protocol Economicalno need to purchase expensive primary and secondary antibodies; considerably less costly than 6xHis-tag antibodies High specificityHisProbe-HRP can recognize His-tags independent of adjacent tags Less steric hindranceHisProbe-HRP conjugate is smaller than antibodies and detects His-tagged fusion proteins that are often undetectable when using some anti-His-tag antibodies78'