b'XCell II Blot Module SureLock Tandem Midi Blot ModuleExpands your XCell SureLock and XCell II Mini-CellThe Invitrogen SureLock Tandem Midi Gel Tank is systems into western transfer devices uniquely designed to enable convenient, reliable gel The Invitrogen XCell II Blot Module allows you toelectrophoresis and protein transfer of high-performance easily transfer proteins or nucleic acids from mini gelsInvitrogen midi gels. With the SureLock Tandem Midi Blot to membranes. It fits neatly into the Invitrogen XCellModule, the tank performs efficient room-temperature SureLock and XCell II Mini-Cell systems in place of thewet transfers for downstream western blot analysis in gel/buffer core assembly. It requires less than 200 mL30 minutes. The tank accommodates two blot modules, of transfer buffer for western, Southern, and northernallowing transfer of one or two gels at a time. The unique transfers. Tough platinized-titanium and stainless-steeldesign of the SureLock Tandem Midi Blot Module uses electrodes create a uniform electrical field without clampsconsiderably less transfer buffer (only ~300 mL per transfer) or hinged gel holders.than other midi-sized wet transfer systems. This lower buffer requirement reduces the amount of hazardous Features: methanol waste.Economicalrequires only 200 mL of methanol-based transfer buffer and helps save you money Features: 2-in-1 midi gel electrophoresis and transfer tankrunFlexiblefits gel sizes up to 8 x 8 cmand transfer high-performance Invitrogen midi gels usingRobust electrodes, sturdy steel platesfor highlythe same tank efficient and reliable western transfersTwo separate gel chambersrun 1 or 2 gels or transfers using only the necessary amount of buffer for Specifications each gel, minimizing buffer cost and wasteMode of transfer WetNuPAGE Bis-Tris and Tris-Acetate, Optimal performance with fast transfer protocolsBolt Bis-Tris Plus, Novex Tris-Glycine,efficient, room-temperature transfers in 30 minutes; Gel compatibility Novex TBE, and Novex TBE-Urea minieliminate the need to prechill buffers and the hassle and gels; pour-your-own gels cast with themessiness of ice bathsSureCast systemRunning dimension VerticalCapacity Up to 2 mini gels SpecificationsGel size Mini (8 x 8 cm) Mode of transfer WetGel compatibility NuPAGE Bis-Tris, NuPAGE Tris-Acetate, and Novex Tris-Glycine Plus midi gelsRecommended products Running dimension VerticalThe PowerEase Touch 350W Power Supply and pre-cutCapacity 1 midi gel per blot modulenitrocellulose and PVDF membranes are recommended forGel size Midi (8 x 13 cm)use with the XCell II Blot Module.View a how-to video on the SureLock Tandem Midi Gel Tank and Blot Module here.Learn more at thermofisher.com/wettransfer9'