b'Membranes for western transfer devices Low-fluorescence membranes for fluorescent Nitrocellulose and PVDF membranes are available aswestern blottingpre-assembled stacks for the Invitrogen iBlot Gel TransferTo eliminate a major source of background fluorescence, Device and Invitrogen Power Blotter System (Tables 5we recommend using membranes with low and 6). The Power Blotter System is also compatible withautofluorescence, including nitrocellulose and specialty pre-cut membranes with filter papers (Table 6). low-fluorescence PVDF membranes (Table 7 and Figure 10). All Invitrogen iBlot 2 and Power Blotter PVDF pre-assembled stacks are equipped with low-fluorescence PVDF membranes for use in fluorescent western blot applications.Table 7. Thermo Scientific and Invitrogen nitrocellulose and PVDF membranes for fluorescent western blotting applications.Mini Midi RollNitrocellulose Nitrocellulose/Filter Paper Sandwiches,Nitrocellulose/Filter Paper Sandwiches,Nitrocellulose Membrane, 0.45 m,0.2 m, 0.2 m, 30 cm x 3.5 m8.3 x 7.3 cm 8.5 x 13.5 cm (Cat. No. 88018)(Cat. No. LC2000) (Cat. No. LC2009)Nitrocellulose/Filter Paper Sandwiches,Nitrocellulose/Filter Paper Sandwiches, 0.45 m, 0.45 m,NA8.3 x 7.3 cm 8.5 x 13.5 cm(Cat. No. LC2001) (Cat. No. LC2006)Low-fluorescenceLow-Fluorescence PVDF Transfer PVDF Membrane, 0.2 m, 7 x 8.4 cm NA NA(Cat. No. 22860)Membrane 488 nm 680 nm 800 nmNitrocellulose, 0.45 mLow- uorescence PVDF, 0.2 mPVDF, 0.45 mFigure 10. Background fluorescence of common membranes.Learn more at thermofisher.com/membranes19'