b'iBright Imaging Systems offer up to five imaging modes to support your multiple applications (Table 24). Efficiently and easily capture data from protein gels, nucleic acid gels, chemiluminescent western blots, fluorescent western blots, and more (Figure 44).Table 24. Imaging modes available on iBright Imaging Systems.Imaging mode What kind of signal can be captured?Colorimetric staining of gels (e.g., Coomassie, silver) and membranes (e.g., Ponceau S, Protein gel Thermo Scientific Pierce Reversible Protein Stain), fluorescent staining of gels (e.g., Invitrogen SYPRO Ruby stain)Nucleic acid gel Ethidium bromide and Invitrogen SYBR stainsChemiluminescence using all popular HRP and AP substrates (e.g., Thermo Scientific Chemiluminescent blot SuperSignal and Invitrogen WesternBreeze substrates) Fluorescence with popular RGB (visible range) and near-IR fluorophores (e.g., Invitrogen Fluorescent blotAlexaFluor and AlexaFluor Plus conjugates)Custom mode to image samples containing multiple signals, such as chemiluminescence, Universalfluorescence, colorimetric stains, and/or visible images; image display is similar to fluorescent blot mode and allows one to assign false color to any sampleFluorescent western blots ChemiluminescentCombined fluorescentColorimetric westernColorimetric stained western blots and chemiluminescentblots protein gelswestern blotsFluorescent stainedFluorescent stainedColorimetric membraneColony plates nucleic acid gels protein gelsstainsFigure 44. Example imaging applications. Images pictured for fluorescent stained nucleic acid gels and colorimetric stained protein gels shown in pseudocolor (false color applied). Data is captured in grayscale.65'