b'A NuPAGE Tris-Acetate gelsB NuPAGE Tris-Acetate gelsRecommended products(denaturing separation) (native separation) Invitrogen HiMark Unstained and Prestained Protein Standards are specifically designed for large protein analysis on NuPAGE Tris-Acetate gels under denaturing conditions. Both standards offer a ready-to-load format and consist of 9 proteins with a size range of 40500 kDa.Figure 15. Migration patterns in NuPAGE Tris-Acetate gels. For optimal results, protein bands should migrate within the gray shaded areas. (A) Migration pattern of Invitrogen HiMark Unstained Protein Standard on a NuPAGE Tris-Acetate gel under denaturing conditions. (B) Migration pattern of Invitrogen NativeMark Unstained Protein Standard on a NuPAGE Tris-Acetate gel under native conditions.Review the quick reference protocol for NuPAGE Tris-acetate mini gels here. Learn more about NuPAGE Tris-Acetate at thermofisher.com/trisacetate23'