b'PowerEase Touch Power SuppliesEasy touchscreen programming and operationThe Invitrogen PowerEase Touch Power Supplies make setting up custom protocols or selecting one of the several preprogrammed gel electrophoresis and transfer methods a breeze with an improved 4.3-inch backlit LCD touchscreen display and user interface. The power supplies are ideal for DNA or RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch Power Supplies offer four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency. The sturdy polyurethane feet and stackable housing designThe PowerEase Touch 120W Power Supply is a medium-allow stacking of power supplies for a reduced footprint onthroughput power supply, with a maximum output of 300 V, the lab bench. 500 mA, and 120 W. The PowerEase Touch 350W Power Supply is a high-throughput power supply, with a maximumEase of useLCD touchscreen display and useroutput of 300 V, 3 A (3,000 mA), and 350 W.interface show clear menu prompts for easy use by hand or stylus and convenient monitoring of run progress Convenientfour sets of output terminals allow running of multiple electrophoresis unitsCustomizableprogram up to 100 custom methods, 20 steps per method, 999 minutes per step, or select one of several preprogrammed Invitrogen gel electrophoresis and transfer methods Safetyfeatures automatic No Load, Over Temperature, Over Voltage, Over Current, Load Change, and Ground Leak detectionLearn more at thermofisher.com/powerease61'