b'Mini Gel TankOne tank, 181 gelsThe Mini Gel Tank is designed for more intuitive use and greater convenience compared to traditional electrophoresis tanks. The unique, side-by-side tank design allows you to perform electrophoresis of 1 or 2 mini gels.The Mini Gel Tank offers:Versatilitycompatible with all of our mini gels, including Invitrogen NuPAGE, Novex, Bolt, and specialty gelsEasy sample loadingforward-facing well configuration Specifications Simultaneous visualization of both gelsstreamlined, Gel capacity: up to 2 mini gelsside-by-side tank configurationCell size (L x W x H): 32 x 11.5 x 16 cm (height with lid on) Simple monitoring of gelswhite tank stand provides Buffer requirement: 400 mL for each mini gel chambereasy visualization of prestained markers Material: polycarbonate Less running buffer requiredgel chambers are separated, so you only need to load sufficient buffer for Chemical resistance: not compatible with acetone, each gel to the specified fill line chlorinated hydrocarbons (e.g., chloroform), or aromatic hydrocarbons (e.g., toluene, benzene)Watch our Mini Gel Tank video at thermofisher.com/minigeltank.Learn more at thermofisher.com/minigeltank54'