b'IEF and specialty protein ladders Protein ladders are available for use in isoelectric focusing (IEF) and for use when the experiment involves the detection of His-tagged, phosphorylated, or glycosylated proteins.IEF His-tagged proteins Phosphorylated proteins Glycosylated proteinspl kDa10.71169.5668.38.0457.87.46.924Novex Gel: Invitrogen pH 310 IEF Protein Gel6.05.35.24.5184.23.5 SimplyBlueInVision 14stain stainInvitrogen IEF Marker 310 Invitrogen BenchMark His-TaggedInvitrogen PeppermintStickInvitrogen CandyCane Protein Standard Phosphoprotein MolecularGlycoprotein Molecular Weight Standards Weight StandardsCat. No. 39212-01 Cat. No. LC5606 Cat. No. P33350 Cat. No. C21852Uses: IEF applications Uses: positive control andUses: positive and negativeUses: positive and negative for molecular weight sizing incontrols for detection ofcontrols for detection of His-tagged fusion protein detection phosphorylated proteins glycosylated proteins Visualization: colorimetric stainVisualization: Invitrogen InVisionVisualization: colorimetricVisualization: colorimetric stain or His-Tag In-Gel Stain or Invitrogenstain or methods that detectmethods that detect glycosylated 6x-His Tag Monoclonal Antibody phosphorylated proteins, suchproteins, such as Invitrogen Pro-Q as Invitrogen Pro-Q DiamondEmerald glycoprotein stain kitsphosphoprotein gel stains51'