b'Running gelsMini gel running conditions in Invitrogen mini gel electrophoresis chamber systems.Voltage (V) Approximate run time (min)Bolt 412% Bis-Tris Plus gel (MES buffer) 200 20Bolt 412% Bis-Tris Plus gel (MOPS buffer) 200 35NuPAGE 412% Bis-Tris gel (MES buffer) 200 30NuPAGE 412% Bis-Tris gel (MOPS buffer) 200 42Novex Tris-Glycine gel, WedgeWell format (denatured) 225 2540Novex Tris-Glycine gel, WedgeWell format (native) 125 6090NuPAGE 38% Tris-Acetate gel (denatured) 150 50NuPAGE 38% Tris-Acetate gel (native) 150 100Novex 1020% Tricine gel 125 65NativePAGE 312% gel 150 80100 60Novex IEF gel, pH 310 200 60500 30Novex 10% Zymogram Plus gel (gelatin) 125 90Note: Run times may vary depending on the power supply and gel percentage.Midi gel running conditions in Invitrogen midi gel electrophoresis chamber systems.Voltage (V) Approximate run time (min)NuPAGE Bis-Tris gel (MES buffer) 200 30NuPAGE Bis-Tris gel (MOPS buffer) 200 40Novex Tris-Glycine Plus gel (denatured) 200 60Novex Tris-Glycine Plus gel (native) 125 120NuPAGE Tris-Acetate gel (denatured) 150 60NuPAGE Tris-Acetate gel (native) 150 135Note: Run times may vary depending on the power supply and gel percentage.62'