b'Buffer recipesBolt Bis-Tris buffer recipesBufferStorageComponentConcentration (1X) Bolt LDS Sample Buffer 425C Glycerol 10%Tris base 141 mMTris-HCl 106 mMLDS 2%EDTA 0.51 mMSERVA Blue G-250 0.22 mMPhenol red 0.175 mM(pH 8.5)Bolt MOPS SDS Running Buffer 425C MOPS 50 mMTris base 50 mMSDS 0.1%EDTA 1 mM(pH 7.7)Bolt MES SDS Running Buffer 425C MES 50 mMTris base 50 mMSDS 0.1%EDTA 1 mM(pH 7.3)Bolt Transfer Buffer 425C Bicine 25 mMBis-Tris (free base) 25 mMEDTA 1.0 mMChlorobutanol 0.05 mM(pH 7.2)43'