b'XCell SureLock Mini-CellSimultaneous electrophoresis of up to 2 mini gelsThe unique design of the Invitrogen XCell SureLock Mini-Cell allows you to run mini gels quickly and easily without any clamps or grease (Figure 27). The tight seal provided by the gel tension wedge results in consistent performance. The XCell SureLock Mini-Cell is compatible with Bolt, NuPAGE, Novex, and specialty gels (Figure 28).Key features of the XCell SureLock Mini-Cell:User-friendly designuses single gel tension wedge with no clamps or greaseFlexibilityperform electrophoresis of 2 mini gels simultaneously Unique, heat dissipating designno need for aSpecificationscooling deviceGel capacity: up to 2 mini gels Built-in usability featuresretractable plugs, recessed Cell size (L x W x H): 14 x 13 x 16 cm (height with lid on)jacks, and a specially designed lid enhances user safetyBuffer chamber requirement (Invitrogen mini gels):Upper buffer chamber: 200 mL Lower buffer chamber: 600 mL Chemical resistance: impervious to most alcohols but not compatible with acetone, chlorinated hydrocarbons (e.g., chloroform), or aromatic hydrocarbons (e.g., toluene, benzene)Learn more at thermofisher.com/surelockmini56'