b'SureCast gel handcast reagentsSureCast Stacking Buffer and Resolving BufferInvitrogen SureCast Stacking Buffer and Resolving Buffer are pouches of dry-blend powder, each sufficient to make 500 mL of stacking gel buffer (0.5 M Tris-HCl buffer, pH 6.8) or 500 mL of resolving gel buffer (1.5 M Tris buffer, pH 8.8) for handcasting polyacrylamide gels. Benefits include: Convenient pouches of dry-blend powderdissolve contents of a single packet in water and the buffer is ready to use Time- and space-savingno weighing, noSee the SureCast Gel Handcast System in calculations, no pH adjustment, and no need to stockaction at thermofisher.com/surecastindividual components Long shelf lifeeasy stocking and storage, as dry powder minimizes concerns about long-term stability of stock solutionsSureCast Acrylamide Solution, 40%Invitrogen SureCast Acrylamide Solution (40%) can be used to prepare single-percentage and gradient gels using the SureCast Gel Handcast System or other handcast systems.Features include: Room-temperature storage Long shelf life High purity Safer alternative to powdered acrylamide Concentrated to enable a broader range of gel percentages to cast35'