b'Fluorescent protein gel stainsRapid, highly sensitive fluorescent stains for total protein detection after electrophoresisFluorescent gel stains are designed for use in 1D and 2D PAGE and offer sensitivities similar to those of silver staining techniques. Invitrogen SYPRO protein stains are easy-to-use fluorescent stains for the detection of proteins separated by PAGE. Stained proteins can be viewed with a standard UV or blue-light transilluminator or with a laser scanner.Features: Simpleno destaining or timed steps required; minimal hands-on time Quantitativelinear quantitation range over two orders of magnitude with low protein-to-protein variability Highly sensitivetypically more sensitive than Coomassie dyebased stains and equivalent to silver stainsSYPRO protein stains.SYPRO Ruby stain SYPRO Orange stain SYPRO Red stainLimit of detection 0.25 ng 48 ng 48 ngStain and destain time 90 min microwave; 18 hr standard ~1 hr ~1 hrEx/Em 280 nm, 450/610 nm 300 nm, 470/510 nm 300 nm, 550/630 nmEase of use Ready to use Supplied as stock solution Supplied as stock solutionCompatible applications Mass spectrometry, IEF, 2D gels,Mass spectrometry, IEF, 2D gels,Mass spectrometry, IEF, 2D gels, on-membrane staining on-membrane staining on-membrane stainingLearn more at thermofisher.com/fluorescentstains70'