b'NativePAGE gelsDid you knowThe blue native polyacrylamide gel electrophoresis technique was developed by Hermann Schgger and Gebhard von Jagow in 1991.Figure 19. NativePAGE gel migration chart. Migration patterns of theRecommended productsNativeMark Unstained Protein Standard on NativePAGE gels are shown. The NativeMark Unstained Protein Standard is recommended for use with native gel chemistries, including our Tris-glycine, Tris-acetate, and NativePAGE gel systems. This standard offers a wide molecular weight range of 201,200 kDa, and the 242 kDa -phycoerythrin is visible as a red band after electrophoresis for reference (prior to staining).27'