b'XCell4 SureLock Midi-CellSimultaneous electrophoresis of up to 4 midi gelsThe Invitrogen XCell4 SureLock Midi-Cell allows simultaneous electrophoresis of 14 midi gels without leaking, enabling consistent performance. The system is designed to dissipate heat effectively and evenly, and enable high-resolution results when using Invitrogen Novex midi gels (Figures 30 and 31).Key features of the XCell4 SureLock Midi-Cell:User-friendly designelectrophoresis without clampsSpecificationsor grease Gel capacity: up to 4 midi gels (8 x 13 cm) Flexibilityperform electrophoresis of 14 midi gelsCell size (L x W x H): 21 x 19 x 16 cm (height with lid on) Unique, heat-dissipating designno need for a Buffer chamber requirement: cooling device Upper buffer chamber: 175 mL x 4 Built-in safety featuresspecially designed lid enhances safetyLower buffer chamber: 540700 mL Chemical resistance: not compatible with acetone, chlorinated hydrocarbons (e.g., chloroform), or aromatic hydrocarbons (e.g., toluene, benzene)Learn more at thermofisher.com/surelockmidi59'