b'Electrophoresis chamber system selection guideOur electrophoresis chamber systems are designed for compatibility with the full range of Invitrogen gel offerings. Refer to the table below to find which system is right for you.XCell SureLock SureLock Tandem XCell4 SureLockMini Gel Tank Mini-Cell Midi Gel Tank Midi-CellGel capacity Up to 2 mini gels Up to 2 mini gels Up to 2 midi gels Up to 4 midi gelsCell dimensions32 x 11.5 x 16 cm14 x 13 x 16 cm 2517.917.3 cm 21 x 19 x 16 cm(L x W x H; height with lid on)AdvantagesThe Mini Gel Tank is Instrument incorporates a Easy-to-use apparatus, with Advanced apparatus versatile and compatiblegel tension wedge in placeseparate chambers for eachfor easier, more reliable with all Invitrogen precastof the rear wedge used ongel, enabling scalable bufferelectrophoresis with and handcast mini gels;earlier models usage midi gelsthe unique tank design XCell II Blot Module is SureLock Tandem Midi Blot enables convenientavailable for semi-wetModule is available for wet side-by-side gel loadingprotein transfers protein transfersand enhanced viewing during use Mini Blot Module is available for wet protein transfersLearn more at thermofisher.com/electrophoresischambers53'