b'Special packaging optionsOptimized kitsWeve packaged many of our popular fluorophores, biotinylation reagents, and crosslinkers in easy-to-use optimized kits for labeling and crosslinking. Each kit contains everything needed to label a protein or antibody with the desired label including the reagents, conjugation buffer, purification system, and an easy-to-follow, step-by-step protocol for conjugation.Highlights:Scalablekits are designed for labeling as little as 100 g of protein and up to 10 mgEasyeverything included with an optimized workflow for labelingWide selectionkits are available for labeling with Alexa Fluor dyes, biotin, and crosslinkersLearn more at thermofisher.com/labelingkitsNo-Weigh packaging formatBioconjugation reagents are highly reactive molecules and therefore may be sensitive to water, light, oxygen, or other surrounding conditions. They may form unstable intermediates that lead to undesired side products of the intended reactions. Therefore, it is critical to minimize the exposure of bioconjugation reagents to these negative environmental factors to obtain the highest yields and quality possible.To mitigate these risks, we offer specific packaging and quality grades for your bioconjugation reagents. In addition to these options, our bioconjugation reagents are available in multiple pack sizes. Custom packaging and pack sizes are also available by request.With the convenient Thermo Scientific No-Weigh format, aHighlights:ready-to-use solution can be made quickly and conveniently. ThisHelps save timeavoid weighing chemicals; just add water, unique packaging format helps to eliminate the need to weigh outbuffer, or solvent to create a working solution in secondssmall volumes of dry chemicals. Once reconstituted, the reagentHelps reduce wastesmall working aliquots limit the amount is ready to use at the desired concentration. Avoid weighingof unused material discardedhassles and wasting precious reagents with our single-use No-Weigh packaging format. Always freshworking solution is ready to use at desired concentration; no need to store stock solutionsLearn more at thermofisher.com/no-weigh108 Protein bioconjugation, crosslinking, and modification thermofisher.com/proteinbiology'