b'Protease and phosphatase inhibitorsBroad-spectrum liquid cocktails and tablets for complete protein protectionProtease and phosphatase inhibitor cocktails and tablets are ideal for the protection of proteins during extraction or lysate preparation from primary cells, cultured mammalian cells, animal tissues, plant tissues, yeast cells, or bacterial cells. Formulations are packaged in multiple sizes, and EDTA-free versions are available for assays that are sensitive to divalent cations.Highlights:Convenientready-to-use, fully disclosed, broad-spectrum formulations available as either liquid cocktails or tablets in multiple pack sizes and with a minimum one year shelf lifeComplete protectioncombined cocktail available with all-in-one formulations containing both protease and phosphatase inhibitors (Table 4)Compatibleuse directly with Thermo Scientific Pierce cell lysis buffers or other commercial or homemade detergent-based lysis reagentsTable 4. Components present in Thermo Scientific Halt Inhibitor Cocktails and Thermo Scientific Pierce Protease and Phosphatase Inhibitor Tablets.Combined protease Protease liquid Phosphatase liquidand phosphatase liquid Inhibitor component Target (mechanism) cocktails and tablets cocktails and tablets cocktails and tabletsAEBSFHCl Serine proteases (irreversible) Aprotinin Serine protease (reversible)Bestatin Aminopeptidase (reversible)E-64 Cysteine (irreversible)Leupeptin Serine and cysteine proteases (reversible)Pepstatin Aspartic acid proteases (reversible) EDTA* Metalloproteases (reversible)Sodium fluoride Serine/threonine and acidic phosphatasesSodium orthovanadate Tyrosine and alkaline phosphatases-Glycerophosphate Serine/threonine phosphataseSodium pyrophosphate Serine/threonine phosphatase* EDTA is not in EDTA-free formulations.For more information, or to view additional products, go to thermofisher.com/inhibitorcocktailsProtein sample preparation thermofisher.com/proteinbiology 17'