b'SureCast Gel Handcast StationPour your own gelsThe Invitrogen SureCast Gel Handcast Station is designed for 100% leak-free protein gel casting. The SureCast system is fully compatible with our Mini Gel Tank.Benefits of the SureCast Gel Handcast System include:Leak-free designgels that are more usable; less wasted timeSuperior glass plate durabilityup to 20 times more durable than other suppliers platesUnique tilt featurehelps minimize spillage when pouring acrylamide solutionsSimple assembly of casting componentsuses a single-motion, load-and-lock mechanismUse Invitrogen SureCast handcasting reagents as well as other popular polyacrylamide gel casting reagents.Bolt Bis-Tris Plus and NuPAGE Bis-Tris mini gelsNeutral-pH gel systems for optimal separationInvitrogen Bolt Bis-Tris Plus and NuPAGE Bis-Tris gels are precast polyacrylamide gels designed for optimal separation of a broad molecular weight range of proteins under denaturing conditions during gel electrophoresis. These gels help deliver consistent performance with a neutral-pH environment to minimize protein degradation, resulting in sharper bands without typical gel smiling. Additionally, preserving protein integrity becomes particularly important when separating low-abundance proteins. The unique wedge-well design of Bolt Bis-Tris Plus gels allows loading up to twice the sample volume of other precast gels. In addition, Bolt gels can be run in as little as 20 minutes. Bolt Bis-Tris Plus and NuPAGE Bis-Tris gels are ideal for western blot transfer and analysis along with any other technique where protein integrity is crucial. Bolt Bis-Tris Plus gels are available in the mini-gel format and NuPAGE Bis-Tris gels are available in mini- and midi-gel formats, as well as multiple thicknesses.Protein gel electrophoresis thermofisher.com/proteinbiology 45'