b'iBright Imaging SystemsExperience an easier time capturing and analyzing data from gels and western blots with Invitrogen iBright Imaging Systems. Designed with a streamlined, intuitive interface and workflows, the iBright Imaging Systems family of instruments are easy to use for researchers of all experience levels.There are three models in the iBright Imaging Systems family: the iBright CL750 Imaging System, the iBright CL1500 Imaging System, and the iBright FL1500 Imaging System. The iBright CL750 Imaging System offers the core essential western blot and gel imaging functions and makes the transition from the darkroom and film easy. The iBright CL1500 Imaging System expands application support and has many of the high-performance specifications of our premier iBright FL1500 model. The iBright FL1500 Imaging System features maximum application support, including fluorescent western blot imagingFive-channel fluorescent blottingmultiplex with the five with up to four fluorescence channels at a time. fluorescence channels of the iBright FL1500 model; capture Highlights: up to four proteins in a single blot for more meaningful and representative experimentsPush-button optimized exposureSmart Exposure acquisition technology for rapid determination of optimalCompliance supportall models offered with 21 CFR Part 11 exposure times helps minimize the need to repeat exposures tocompliance support software packages to set up and control get the desired signalsecurity, audit, and e-signature settingsPowerful 9.1 megapixel (MP) cameracapture crystal-cleariBright Imaging Systems offer up to five imaging modes images with robust imaging potential to support multiple applications (Table 3). Efficiently and Advanced automated featuresautomatic sample rotation,*easily capture data from protein gels, nucleic acid gels, automatic zoom, automatic focus, and automatic onboard datachemiluminescent western blots, fluorescent western blots, and analysis provide a smooth imaging experience more (Figure 8).* Automatic sample rotation available on CL1500 and FL1500 models.Invitrogen iBright CL750 Invitrogen iBright CL1500 Invitrogen iBright FL1500Imaging System Imaging System Imaging SystemCore essential western blot Expanded application support Maximum application support, and gel imaging functions to efficientlywith many of the sameincluding fluorescent westerntransition from the darkroom and film high-performance specifications as theblot imaging with up to four premier iBright FL1500 model fluorescence channels at a time70 Western blotting thermofisher.com/proteinbiology'