b'SureLock Tandem Midi Blot Module The Invitrogen SureLock Tandem Midi Gel Tank is uniquely designed to enable convenient, reliable gel electrophoresis and protein transfer using high-performance Invitrogen midi gels. With the SureLock Tandem Midi Blot Module, the tank performs efficient room temperature wet transfers for downstream western blot analysis in 30 minutes. The tank accommodates two blot modules, allowing transfer of one or two gels at a time. The unique design of the SureLock Tandem Midi Blot Module uses considerably less transfer buffer (only ~300 mL per transfer) than other midi-size wet transfer systems. This lower buffer requirement reduces the amount of hazardous methanol waste.Highlights:2-in-1 midi gel electrophoresis and transfer tankseparate and transfer proteins in the same tank using high-performance Invitrogen midi gelsTwo separate gel chambersrun 1 or 2 gels or transfers using only the necessary amount of buffer for each gel, minimizing buffer cost and wasteOptimal performance with fast transfer protocolsefficient, room temperature transfers in 30 minutes; eliminate the need to prechill buffers and the hassle and messiness of ice bathsView the how-to video:Learn more at thermofisher.com/wettransferWestern blotting thermofisher.com/proteinbiology 61'