b'ProcartaPlex multiplex immunoassay kitsProfile more biomarkers with less sampleMore than 90% of ProcartaPlex assays can be combined with ProcartaPlex multiplex immunoassays use the Luminex xMAPone another(multi-analyte profiling) technology that enables the simultaneousAll ProcartaPlex assays are individually tested for specificity detection and quantitation of up to 80 protein targets in aand endogenous, native protein detectionsingle 2550 L samplefrom plasma, serum, cell cultureMulti-analyte detection with a large dynamic rangesupernatants, and other bodily fluids. The technology employs the use of differentially dyed capture beads for each target inScalable and reproducible performance regardless of plex sizea multiplex ELISA-like assay. ProcartaPlex immunoassays canLargest multiplex panel on the market for quantitation of up to profile up to 80 times more analytes using significantly less80 analytessample in the same time that it takes to perform a traditionalProcartaPlex multiplex immunoassays are available in multiple sandwich ELISA. formats across six species (human, mouse, rat, nonhuman ProcartaPlex multiplex immunoassays are available in off-the- primate, porcine, and canine) to meet the needs of your research.shelf kits or you can create your own unique panel through ourGet information regarding characteristic assay details and mix-and-match panel offering (Table 6). It is often advantageousindividual analytes at thermofisher.com/procartaplexto start with a large panel and then reduce the number of biomarkers analyzed in a more focused panel. Whatever option works best for you, ProcartaPlex panels enable reproducible results throughout the course of your study. Table 6. ProcartaPlex multiplex assay options.MixingBeadBase kit Name Description required type includedProcartaPlexPredefined, biologically relevant, and disease-defined panels using Preconfiguredmagnetic beads for the quantitative multiplex analysis of up to 80 analytesNo Magnetic Yespanels in a single sample. Optimal performance and reproducibility through extensive testing for combinability, cross-reactivity, and interference.ProcartaPlexCustom-blended and optimized panels deliver results tailored to the panel Mix & Matchdesign of your choice. Simply provide your desired species, sample type,No Magnetic Yespanels and instrument for use. Then select your desired analytes, and we will build and optimize a custom assay kit according to your specifications.Bead sets for the detection of individual analytes designed to be added ProcartaPlexto ProcartaPlex panels for increased customization. Alternatively, multiple ProcartaPlex simplex bead sets can be combined and run using theYes Magnetic NoSimplex sets Invitrogen ProcartaPlex Basic Kit, which includes all nontarget-specific reagents needed to perform the assay.Have technical questions? Need help getting started?Email luminexfas@thermofisher.com to get a one-on-one technical consultation.Immunoassays thermofisher.com/proteinbiology 83'