b'Protein Expression Basics online learning hubGibco Protein Expression Basics is an online hub that brings together educational content and tools to enhance your research. Here you can access our protein expression selection guide, handbook, educational videos, and technical resources. Start here if you are new to protein expression or need to refresh your knowledge. Access the learning hub at thermofisher.com/gibcoproteinexpressionbasicsProtein sample preparationProtein assay technical handbookDownload this handbook to help select the appropriate protein assay method based on the assay time, sensitivity, compatibility, standard curve linearity, and protein-to-protein variation. Learn about our wide range of colorimetric (copper- or dye-based) and fluorescence protein assays, as well as our more specialized assays to quantifypeptides, antibodies, protein modifications, or functional (enzymatic) classes of proteins.Protein assay technical handbook Discover tools and strategies to help optimize your protein quantitation assays to helpTools and reagents for improved quantitation oftotal or specific proteinsensure more accurate downstream results. Download the free handbook at thermofisher.com/protein-assay-handbookProtein clean-up technical handbookLearn how to effectively remove contaminants, perform buffer exchange, or concentrate protein samples from 2 L to 250 mL using various Thermo Scientific protein biology tools in this 48-page handbook. Dialyze protein samples securely using Slide-A-Lyzer cassettes and devices. Rapidly desalt samples with high protein recovery using Zeba desalting spin columns and plates. Efficiently extract specific contaminants using resins optimized for detergent or endotoxin removal. Concentrate dilute protein samples quickly using Pierce Protein Concentrators.Download the free handbook at thermofisher.com/proteincleanuphandbookProtein Sample Preparation eLearning CourseThis free course comprises two interactive modules that were developed to provide a succinct, contextual overview of protein sample isolation using total protein extraction and subcellular fractionation. The module covers methods for purifying whole cell lysates and describes strategies for protein cleanup procedures. Access the free course at thermofisher.com/elearningproteinsampleprepResources thermofisher.com/proteinbiology 121'