b'Wet transfer systemsMini Blot ModuleThe Invitrogen Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank and is designed to make your western blot transfers simple and easy to perform. The tank accommodates one Mini Blot Module per chamber, or two blot modules total in the side-by-side layout. The universal connection and molded gasket make the blot module easy to use, while the inner core of the blot module allows for use of less methanol-based transfer buffer per wet transfer than other commercially available transfer systems. At the recommended conditions and constant voltage, proteins can typically be transferred to nitrocellulose or PVDF membranes in 30 to 60 minutes.Highlights:Universal module designallows modules to fit in either chamber of the tank, simplifying the transfer setupUnique gasket sealhelps prevent buffer leakage to minimize mess during setup of your western blot transfer1/2-inch buffer chamberrequires only 220 mL per blot of methanol-based transfer buffer, helping save you money on buffer and disposal costsStandard 60-minute transfer protocolaccelerates your workflow so you can get results fasterRobust electrodes, sturdy steel platesfor highly efficient and reliable western blot transfersView the how-to video:60 Western blotting thermofisher.com/proteinbiology'