b'Zeba desalting productsConvenient spin formats help ensure rapid desalting with high protein recoveryThermo Scientific Zeba desalting products contain proprietary high-performance resins with exceptional desalting and protein recovery characteristics. They can help process even very dilute protein samples with high levels of protein recovery and greater than 95% retention (removal) of salts and other small molecules. The resin is provided in convenient spin columns, plates, and cartridges, for processing sample volumes between 2 L and 4 mL (Tables 68).Highlights:High performanceproprietary resin enables excellent protein recovery and efficient contaminant removal (Figure 6)Flexibleavailable in spin columns, filter spin plates, and cartridges for a range of needsFastno fraction screening or waiting for protein to emerge by gravity flowEconomicalcost-effective products that offer great performanceTable 6. Zeba desalting products selection guide by format and recommended sample volume.Type Spin columns Spin plate Chromatography columnsFormat Micro 0.5 mL 2 mL 5 mL 10 mL 96-well 1 mL 5 mLResin bed 75 L 0.5 mL 2 mL 5 mL 10 mL 550 L 1 mL 5 mLSample volume 212 L 30130 L 200700 L 5002,000 L 7004,000 L 20100 L 50250 L 1001,500 L(7K MWCO)Sample volume514 L 70200 L 200900 L 3002,000 L 1,0004,000 L 20100 L NA NA(40K MWCO)20 Protein sample preparation thermofisher.com/proteinbiology'