b'Recombinant protein purificationWe offer a variety of resins for the purification of recombinant proteins. These resins are available in multiple formats to accommodate a variety of needs, from high-throughput screening to batch and pilot-scale purification. Superflow resins have undergone extensive chemical characterization. We have ligands targeting a variety of fusion tags, including 6xHis, GST, FLAG epitope, c-Myc, and HA (Table 5).Table 5. Thermo Scientific recombinant protein purification selection guide.High-throughputHigh-throughput Tag Ligand Features Recommended product screening batch Batch PilotDYKDDDDK ImmobilizedPierce Anti-DYKDDDDK Magnetic Agarose (FLAG) Anti-FLAG antibody Pierce Anti-DYKDDDDK Affinity Resin (UltraLink resin)c-Myc Antic-Myc ImmobilizedPierce Antic-Myc Magnetic Beads antibody Pierce Antic-Myc Agarose (Superflow)HA Anti-HA ImmobilizedPierce Anti-HA Magnetic Beads antibody Pierce Anti-HA Agarose Pierce Ni-NTA Magnetic Agarose Beads Ni-NTA orHigher proteinProBond Nickel Chelating Resin Ni-IDA yield HisPur Ni-NTA Agarose Resin 6xHis HisPur Ni-NTA Superflow Resin Higher proteinDynabeads His-Tag Isolation Magnetic Beads Cobalt purity HisPur Cobalt Agarose Resin HisPur Cobalt Superflow Resin Solubility andPierce Glutathione Magnetic Agarose Beads GST Glutathione purification tag Pierce Glutathione Agarose Pierce Glutathione Superflow Agarose For more information, or to view additional products, go to thermofisher.com/proteinpurification34 Protein purification thermofisher.com/proteinbiology'