b'PowerEase Touch Power SuppliesEasy touchscreen programming and operation The Invitrogen PowerEase Touch Power Supplies make setting up custom protocols or selecting one of the several preprogrammed gel electrophoresis and transfer methods a breeze with an improved 4.3-inch backlit LCD touchscreen display and user interface. The power supplies are ideal for DNA or RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch Power Supplies offer four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency. The sturdy polyurethane feet and stackable housing design allow stacking of power supplies for a reduced footprint on the lab bench.The PowerEase Touch 120W Power Supply is a medium-Ease of useLCD touchscreen display and user interfacethroughput power supply, with a maximum output of 300 V, show clear menu prompts for easy use by hand or stylus and500 mA, and 120 W. The PowerEase Touch 350W Power Supply convenient monitoring of run progressis a high-throughput power supply, with a maximum output of Convenientfour sets of output terminals allow running of300 V, 3 A (3,000 mA), and 350 W.multiple electrophoresis units Customizableprogram up to 100 custom methods, 20 steps per method, 999 minutes per step, or select one of several preprogrammed Invitrogen gel electrophoresis and transfer methodsSafetyfeatures automatic No Load, Over Temperature, Over Voltage, Over Current, Load Change, and Ground Leak detectionProtein gel electrophoresis thermofisher.com/proteinbiology 51'