b'Uncoated ELISA and antibody pair kitsCoat it yourself: optimized reagents, all in one box 3.5Uncoated ELISA and antibody pair kits are ideal for experienced ELISA users on a budget, who have less stringent requirements3.0for inter- and intra-assay variance. Affordable ELISA kits using Absorption (450 nm)2.5coat-it-yourself plates also include the necessary reagents to prepare and run an ELISA, including ELISA-optimized matched2.0antibody pairs, standards, detection reagents, wash buffers, and coating buffers (plates are optional). 1.5Highlights: 1.0Flexiblepurchase based on the volume of your assay0.5through a choice of package sizes, including 2-, 10-, and 20-plate kits 0.00 2 4 6 8 10 12 14Complete and easy to useincludes optimized antibodyConcentration (ng/mL)pairs and recombinant standards (Figure 3), TMB substrate, and all essential reagents and buffersunlike ELISA kits fromFigure 3. Representative standard curve for Invitrogen Mouse other suppliers TGF Beta-1 Uncoated ELISA Kit with Plates.Affordablepriced to accommodate the most demanding budgets and to help maximize your researchFind out more about uncoated ELISA kits at thermofisher.com/uncoatedelisa78 Immunoassays thermofisher.com/proteinbiology'