b'Magnetic standsMultiple formats for low- to high-throughput sample processing Invitrogen DynaMag magnets isolate any target in combination with Dynabeads magnetic beads. Reduce waiting time as these powerful magnets quickly pull the bead-bound target to the tube wall. The DynaMag magnets help to ensure optimal working positions and are functionally adapted to suit various workflows.DynaMag-2 MagnetHighlights:Optimizeddeveloped and certified for use with Dynabeads magnetic beadsEasy to handledesigned with ergonomics in mindMore choicesdifferent formats to accommodate different volume and throughput needsThe Invitrogen DynaMag-2 Magnet is our most commonly used magnet for immunoprecipitation and holds up to 16 standard 1.52 mL tubes in numbered spaces. The optimal working volume is 102,000 L. There are eight positions on each side of the rack. The top rack can be quickly removed from the magnet in the base, ready for vortexing, rotation, or manual sample shaking. A center pin in the rack ensures equal vortexing of all tubes, providing excellent control and visibility during sample processing.DynaMag-Spin MagnetInvitrogen DynaMag-Spin Magnet, which accommodates up to six (1.5 mL) microcentrifuge tubes, supports a working volume of 101,500 L. The circular top rack can be quickly removed from the magnet in the base, ready for vortexing or manual sample shaking. Plate-based magnetic stands, such as the Invitrogen DynaMag-96 series, are ideal for manual and automated work, with a footprint the same size as that of a 96-well plate. The recommended working volume is 5200 L.DynaMag-96 Side MagnetSample rack forDynaMag-96 Bottom MagnetDynaMag-2 MagnetDynaMag-96 Side Skirted MagnetTest your protein research knowledge Question: For large yields of highly purified protein, which purification strategy is the best?A. Immunoprecipitation B. Ion exchange chromatography C. Affinity chromatography D. Co-immunoprecipitationAnswer: C38 Protein purification thermofisher.com/proteinbiology'