b'Bandmate Automated Western Blot ProcessorThe Invitrogen Bandmate Automated Western Blot Processor is a programmable blot rocking system that automates the tedious hands-on blocking, washing, and antibody incubation steps of western blot processing. Minimal effort is required to set up the Bandmate device to process up to 2 midi blots or 4 mini blots using your current optimized reagents and protocols for blot processing, freeing up time for other important tasks.Load-and-go setupprepare block, wash, and antibody solutions, load into the machine, select a program, and walk awayWorks with traditional western blotting protocolsno need to switch from current protocols and no specialized reagents required (Figure 4); program the timing of steps based on preference or use preprogrammed optionsAntibody recoverycollection tubes can recover antibody for reuse in future experiments if desiredFigure 4. Comparison of mini blots processed with the Bandmate Automated Western Blot Processor vs. manual processing (probing and washing steps performed in a tray and on a shaker platform). The blots processed with the Bandmate device show comparable or higher intensity levels across multiple probed targets compared to the blots processed manually.View the how-to video:Learn more at thermofisher.com/bandmateWestern blotting thermofisher.com/proteinbiology 65'